missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenyl-5’-oxobutyric Acid, TRC

Product Code. 30407724
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407724 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30407724 Supplier Toronto Research Chemicals Supplier No. TRCF58849550MG
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Fluorophenyl-5'-oxobutyric Acid
CAS 149437-76-3
Molecular Formula C11 H11 F O3
Quantity 50 mg
Synonym 4-(4-Fluorobenzoyl)-Butyric Acid
InChI Formula InChI=1S/C11H11FO3/c12-9-6-4-8(5-7-9)10(13)2-1-3-11(14)15/h4-7H,1-3H2,(H,14,15)
SMILES OC(=O)CCCC(=O)c1ccc(F)cc1
IUPAC Name 5-(4-fluorophenyl)-5-oxopentanoic acid
Molecular Weight (g/mol) 210.2
Formula Weight 210.0692
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.