missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenyl-5’-oxobutyric Acid, TRC

Product Code. 30394115
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394115 500 mg 500mg
30391018 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394115 Supplier Toronto Research Chemicals Supplier No. TRCF588495500MG
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Fluorophenyl-5'-oxobutyric Acid
CAS 149437-76-3
Molecular Formula C11 H11 F O3
Quantity 500 mg
Synonym 4-(4-Fluorobenzoyl)-Butyric Acid
InChI Formula InChI=1S/C11H11FO3/c12-9-6-4-8(5-7-9)10(13)2-1-3-11(14)15/h4-7H,1-3H2,(H,14,15)
SMILES OC(=O)CCCC(=O)c1ccc(F)cc1
IUPAC Name 5-(4-fluorophenyl)-5-oxopentanoic acid
Molecular Weight (g/mol) 210.2
Formula Weight 210.0692
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.