Learn More
(4-Fluorophenyl)methanesulfonyl chloride, ≥95%, Thermo Scientific™
Product Code. p-3175193
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 10576694
Brand: Thermo Scientific Maybridge MO07801CB
Request Bulk or Custom Format
Chemical Identifiers
CAS 103360-04-9
Molecular Formula C7H6ClFO2S
Molecular Weight (g/mol) 208.631
MDL Number MFCD01631930
InChI Key UUQGWVIRPCRTSA-UHFFFAOYSA-N
Synonym 4-fluorophenyl methanesulfonyl chloride, 4-fluorobenzylsulfonyl chloride, 4-fluoro-phenyl-methanesulfonyl chloride, 4-fluorophenyl methyl sulfonyl chloride, chloro 4-fluorophenyl methyl sulfone, 4-fluorophenyl methanesulphonyl chloride, 4-fluoro-alpha-toluenesulfonyl chloride, benzenemethanesulfonyl chloride, 4-fluoro, 4-fluoro-phenyl-methane sulfonyl chloride, pubchem10056
PubChem CID 2759108
IUPAC Name (4-fluorophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC=C1CS(=O)(=O)Cl)F
Specifications
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 208.64
Percent Purity 95%
Chemical Name or Material (4-Fluorophenyl)methanesulfonyl chloride