Learn More
Justrite™ 4 Gallon Sure-Grip™ EX Countertop Corrosives/Acid Steel Safety Cabinet With 1 Self-Closing Door
Click to view available options
:
4 Gallon, Self-Closing, Blue
Description
Countertop and Compac Cabinets let you customize your work areas and make the most of available space. Whether adding to existing storage space or making chemicals readily available at workstation locations, these smaller-sized cabinets are the solution when space is at a premium.
Specifications
Specifications
| Description | 4 gallon safety cabinet with self-closing door; 18 Ga CR Steel |
| Dimensions (D x W x H) | 17 x 17 x 22 in. |
| Material | Steel (CR) |
| Product Type | Safety Cabinet |