Learn More
4-Heptyn-2-ol, 97%
Product Code. 11357676
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11357676 25 g 25g
11347676 5 g 5g
2 options
Product Code. 11357676
Brand: Thermo Scientific Alfa Aesar L11292.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 19781-81-8 | C7H12O | 112.17 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 19781-81-8
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00041588
InChI Key CXVIEBMEWKSONY-UHFFFAOYNA-N
Synonym 4-heptyn-2-ol, acmc-1c9g1, cxviebmewksony-uhfffaoysa
PubChem CID 89225
IUPAC Name hept-4-yn-2-ol
SMILES CCC#CCC(C)O
Specifications
Density 0.88
Boiling Point 67°C to 68°C (14 mmHg)
Flash Point 62°C (143°F)
Refractive Index 1.4515
Quantity 25 g
Beilstein 2232633
Formula Weight 112.17
Percent Purity 97%
Chemical Name or Material 4-Heptyn-2-ol