Learn More
4-Hydroxy-2-hexenal Antibody (6F10), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59352IR
Product Code. 18229692
  • € 720.00 / 100µL
Description
4-Hydroxy-2-hexenal Monoclonal specifically detects 4-Hydroxy-2-hexenal in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4-Hydroxy-2-hexenal
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
Synthetic 4-Hydroxy-2-hexenal modified Keyhole Limpet Hemocyanin (KLH).
100 μL
Specific for 4-Hydroxy-2-hexenal (4-HHE) and 4-Hydroxynonenal (4-HNE) modified proteins. Does not detect free 4-Hydroxy-2-hexenal (4-HHE) or 4-Hydroxynonenal (4-HNE). Does not cross-react with Acrolein, Crotonaldehyde, Hexanoyl Lysine, Malondialdehyde, or Methylglyoxal modified proteins.
Store at 4C in the dark.
IgG2b
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
6F10
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Mouse
Protein G purified
Primary
Non-species specific