Learn More
Invitrogen™ 4-Hydroxy-2-hexenal Monoclonal Antibody (6F10), PE
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA545788
Product Code. 17838152
  • € 481.00 / 100µg
Description
A 1:1,000 dilution of MA5-45788 was sufficient for detection of 4-Hydroxy-2-hexenal in 0.5 μg of 4-Hydroxy-2-hexenal conjugated to BSA and 4-Hydroxy nonenal conjugated to BSA by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary Antibody. Specific for 4-Hydroxy-2-hexenal (4-HHE) and 4-Hydroxynonenal (4-HNE) modified proteins. Does not detect free 4-Hydroxy-2-hexenal (4-HHE) or 4-Hydroxynonenal (4-HNE). Does not cross-react with Acrolein, Crotonaldehyde, Hexanoyl Lysine, Malondialdehyde, or Methylglyoxal modified proteins.
4-Hydroxynonenal, or 4-hydroxy-2-nonenal or 4-HNE or HNE, (C9H16O2), is an alpha,beta-unsaturated hydroxyalkenal that is produced by lipid peroxidation in cells. 4-HNE is the primary alpha,beta-unsaturated hydroxyalkenal formed in this process.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4-Hydroxy-2-hexenal
Monoclonal
1 mg/mL
2.48mM MES, 95.64mM phosphate with 2mM EDTA and no preservative; pH 7.4
Mouse
Protein G
RUO
Chemical
Antibody
IgG2b
ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
6F10
PE
4-HHE; 4-hydroxy Hexenal; 4-Hydroxy-2-hexenal; 4-Hydroxy-2-hexenal-modified; HHE; MG; MG-modified protein
Synthetic 4-Hydroxy-2-hexenal modified Keyhole Limpet Hemocyanin (KLH).
100 μg
Primary
4°C
Liquid
Chemical