missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-2-hexenal Mouse, Biotin, Clone:6F10, Abnova™
Mouse monoclonal antibody raised against synthetic 4-Hydroxy-2-hexenal (4-HHE).
Brand: Abnova MAB17396.100ug
This item is not returnable.
View return policy
Specifications
| 4-Hydroxy-2-hexenal | |
| Monoclonal | |
| Biotin | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| Synthetic 4-Hydroxy-2-hexenal modified Keyhole Limpet Hemocyanin (KLH). | |
| Store at -20°C. | |
| Liquid |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| 6F10 | |
| ELISA (1:1000) Flow Cytometry (1:50) Immunocytochemistry (1:50) Immunofluorescence (1:50) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
| Mouse | |
| 100 μg | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction