Learn More
(4-Hydroxy-3-methoxyphenyl)acetic-2,2-d2 Acid, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Homovanillic Acid D2 (acetic-2,2-D2 acid) |
| CAS | 53587-33-0 |
| Molecular Formula | C9 D2 H8 O4 |
| Quantity | 0.01 g |
| Synonym | Benzeneacetic-d2 acid, 4-hydroxy-3-methoxy- (9 CI), Homovanillic-2H2 acid, 2-(4-Hydroxy-3-methoxyphenyl)dideuterioacetic acid, 4-Hydroxy-3-methoxybenzeneacetic-d2 acid, 2-(4-Hydroxy-3-methoxyphenyl)acetic-2,2-d2 acid, 3'-Methoxy-4'-hydroxyphenylacetic-2,2-d2 acid, 3'-Methoxy-4'-hydroxyphenylacetic-2,2-d2 acid, 3'-Methoxy-4'-hydroxyphenylacetic-2,2-d2 acid, 3'-Methoxy-4'-hydroxyphenylacetic-2,2-d2 acid, 3'-Methoxy-4'-hydroxyphenylacetic-2,2-d2 acid, 3'-Methoxy-4'-hydroxyphenylacetic-2,2-d2 acid, 3'-Methoxy-4'-hydroxyphenylacetic-2,2-d2 acid |
| InChI Formula | InChI=1 S/C9H10O4/c1-13-8-4-6(5-9(11)12)2-3-7(8)10/h2-4,10 H,5H2,1H3,(H,11,12)/i5D2 |
| SMILES | [2 H]C([2 H])(C(=O)O)c1ccc(O)c(OC)c1 |
| IUPAC Name | 2,2-dideuterio-2-(4-hydroxy-3-methoxyphenyl)acetic acid |
| Molecular Weight (g/mol) | 184.1856 |
| Formula Weight | 184.0705 g/mol |
| Show More |