Learn More
4-Hydroxy-3-nitropyridine, 95%
Product Code. 11470027
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470027 5 g 5g
11480027 25 g 25g
11490027 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11470027 Supplier Thermo Scientific Alfa Aesar Supplier No. B20963.06
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
CAS: 5435-54-1 | C5H4N2O3 | 140.098 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5435-54-1
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.098
MDL Number MFCD00128874
InChI Key YUWOLBZMQDGRFV-UHFFFAOYSA-N
Synonym 4-hydroxy-3-nitropyridine, 3-nitropyridin-4-ol, 3-nitro-4-pyridinol, 3-nitro-4-hydroxypyridine, 3-nitropyridin-4 1h-one, 3-nitro-4-pyridone, 4-pyridinol, 3-nitro, 4 1h-pyridinone, 3-nitro, 3-nitro-1,4-dihydropyridin-4-one, 4-hydroxy-3-nitropytidine
PubChem CID 228506
IUPAC Name 3-nitro-1H-pyridin-4-one
SMILES C1=CNC=C(C1=O)[N+](=O)[O-]
Specifications
Melting Point ∼280°C (decomposition)
Quantity 5 g
Beilstein 1072928
Formula Weight 140.1
Percent Purity 95%
Chemical Name or Material 4-Hydroxy-3-nitropyridine