missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-5-iodopyrimidine, 95%, Thermo Scientific Chemicals

Catalog Number p-7053622
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
4349-07-9
CHEBI:17780
This item is not returnable. View return policy
4349-07-9
CHEBI:17780

4-Hydroxy-5-iodopyrimidine is an intermediate used in pharmaceuticals and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-5-iodopyrimidine is an intermediate used in pharmaceuticals and chemical research.

Solubility
Soluble in water (67 g/L) (25°C).

Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Molecular Formula C11H12N2O3
MDL Number MFCD00064341
Quantity 250 mg
Sensitivity Light sensitive
Synonym 5-hydroxy-l-tryptophan, l-5-hydroxytryptophan, oxitriptan, 5-hydroxytryptophan, levothym, tript-oh, cincofarm, l-5-htp, pretonine, 2s-2-amino-3-5-hydroxy-1h-indol-3-yl propanoic acid
Solubility Information Soluble in water (67g/L) (25°C).
InChI Key LDCYZAJDBXYCGN-IMWMWJONNA-N
SMILES N[C@@H](CC1=CNC2=CC=C(O)C=C12)C(O)=O
IUPAC Name (2S)-2-amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
Molecular Weight (g/mol) 220.23
PubChem CID 439280
Formula Weight 221.98
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Hydroxy-5-iodopyrimidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'