missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-D-(-)-2-phenylglycine, TRC

Product Code. 30134845
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134845 10 g 10g
17974365 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134845 Supplier Toronto Research Chemicals Supplier No. TRCH94925010G
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22818-40-2
Molecular Formula C8 H9 N O3
Molecular Weight (g/mol) 167.16
Synonym (2R)-2-Amino-2-(4-hydroxyphenyl)acetic acid, Cefadroxil Monohydrate Imp. A (EP), Amoxicillin Trihydrate Imp. I (EP), Benzeneacetic acid, α-amino-4-hydroxy-, (αR)-, Benzeneacetic acid, α-amino-4-hydroxy-, (R)-, Glycine, 2-(p-hydroxyphenyl)-, D- (8CI), (αR)-α-Amino-4-hydroxybenzeneacetic acid, (2R)-2-Azaniumyl-2-(4-hydroxyphenyl)acetate, (R)-(4-Hydroxyphenyl)glycine, (R)-2-(4-Hydroxyphenyl)glycine, (R)-2-Amino-2-(4-hydroxyphenyl)acetic acid, 4-Hydroxy-D-phenylglycine, D-(-)-2-(4-Hydroxyphenyl)glycine, D-(-)-2-(p-Hydroxyphenyl)glycine, D-(-)-4-Hydroxyphenylglycine, D-(-)-Amino(4-hydroxyphenyl)acetic acid, D-(-)-p-Hydroxyphenylglycine, D-(-)-α-(4-Hydroxyphenyl)glycine, D-(-)-α-Amino-4-hydroxyphenylacetic acid, D-(-)-α-Amino-p-hydroxyphenylacetic acid, D-2-(4-Hydroxyphenyl)glycine, D-2-Amino-2-(p-hydroxyphenyl)acetic acid, D-4-Hydroxyphenylglycine, D-p-Hydroxyphenylglycine, D-α-Amino-4-hydroxyphenylacetic acid, D-α-Amino-p-hydroxyphenylacetic acid, D-α-p-Hydroxyphenylglycine, p-Hydroxy-(R)-phenylglycine, p-Hydroxy-D-phenylglycine
IUPAC Name (2R)-2-amino-2-(4-hydroxyphenyl)acetic acid
SMILES N[C@@H](C(=O)O)c1ccc(O)cc1

Specifications

InChI Formula InChI=1S/C8H9NO3/c9-7(8(11)12)5-1-3-6(10)4-2-5/h1-4,7,10H,9H2,(H,11,12)/t7-/m1/s1
Quantity 10 g
Formula Weight 167.0582
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material D-alpha-(4-Hydroxy-phenyl)glycine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.