Learn More
Thermo Scientific Chemicals (-)-4-Hydroxy-D-phenylglycine, 98+%
Product Code. 11317176
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317176 25 g 25g
11307176 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11317176 Supplier Thermo Scientific Chemicals Supplier No. L07190.14
 Restricted Item
In Stock
Add to basket
CAS: 22818-40-2 | C8H9NO3 | 167.164 g/mol
(-)-4-Hydroxy-D-phenylglycine is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(-)-4-Hydroxy-D-phenylglycine is used as an organic chemical synthesis intermediate.
Solubility
Soluble in water (5 g/L at 20°C) and 1 mol/l HCl.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22818-40-2
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00004262
InChI Key LJCWONGJFPCTTL-SSDOTTSWSA-N
Synonym d-4-hydroxyphenylglycine, 4-hydroxy-d-phenylglycine, d--4-hydroxyphenylglycine, h-d-phg 4-oh-oh, d-n-4-hydroxyphenyl glycine, 2r-2-amino-2-4-hydroxyphenyl acetic acid, r-2-amino-2-4-hydroxyphenyl acetic acid, r-alpha-amino-4-hydroxybenzeneacetic acid, d---4-hydroxyphenylglycine, unii-pcm9oix717
PubChem CID 89853
ChEBI CHEBI:15695
IUPAC Name (2R)-2-amino-2-(4-hydroxyphenyl)acetic acid
SMILES C1=CC(=CC=C1C(C(=O)O)N)O
Specifications
Melting Point ∽240°C (decomposition)
Beilstein 2210998
Solubility Information Soluble in water (5g/L at 20°C) and 1 mol/l HCl.
Quantity 25 g
Formula Weight 167.16
Optical Rotation −156° (c=1 in 1N HCl)
Percent Purity ≥98%
Chemical Name or Material (-)-4-Hydroxy-D-phenylglycine