missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxybenzamidine, Hydrochloride, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxybenzamidine, Hydrochloride |
| CAS | 38148-63-9 |
| Molecular Formula | C7 H8 N2 O . Cl H |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C7H8N2O.ClH/c8-7(9)5-1-3-6(10)4-2-5;/h1-4,10H,(H3,8,9);1H |
| SMILES | Cl.NC(=N)c1ccc(O)cc1 |
| IUPAC Name | 4-hydroxybenzenecarboximidamide;hydrochloride |
| Molecular Weight (g/mol) | 172.61 |
| Formula Weight | 172.0403 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction