Learn More
4-Hydroxybiphenyl-d9 (rings-d9), TRC
Product Code. 30424847
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424847 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30424847 Supplier Toronto Research Chemicals Supplier No. TRCP3361365MG
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Hydroxybiphenyl ring-d9
CAS 126840-28-6
Molecular Formula C12 2H9 H O
Quantity 5 mg
Synonym [1,1'-Biphenyl-2,2',3,3',4',5,5',6,6'-d9]-4-ol (9CI), p-Phenylphenol (ring-D9), 4-Phenylphenol D9, 4-Hydroxy-1',2,2',3,3',4',5,5',6,6'-nonadeuteriobiphenyl, 4-Hydroxybiphenyl-1',2,2',3,3',4',5,5',6,6'-D9, 4-Hydroxybiphenyl (ring-D9)
InChI Formula InChI=1S/C12H10O/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9,13H/i1D,2D,3D,4D,5D,6D,7D,8D,9D
SMILES [2H]c1c([2H])c([2H])c(c([2H])c1[2H])c2c([2H])c([2H])c(O)c([2H])c2[2H]
IUPAC Name 2,3,5,6-tetradeuterio-4-(2,3,4,5,6-pentadeuteriophenyl)phenol
Molecular Weight (g/mol) 179.26
Formula Weight 179.1297