Learn More
4-Hydroxynonenal Antibody (12F7), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59353B
Product Code. 18227991
  • € 731.00 / 100µL
Description
4-Hydroxynonenal Monoclonal specifically detects 4-Hydroxynonenal in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4-Hydroxynonenal
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
Mouse
Protein G purified
Primary
Non-species specific
Purified
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
12F7
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
(2E)-4-Hydroxy-2-nonenal, 2-Nonenal, 4-hydroxy-, (2E,4R)-, 4 HNE, 4-hydroxy-trans-2-nonenal
Synthetic 4-Hydroxynonenal modified Keyhole Limpet Hemocyanin (KLH)
100 μL
Specific for 4-Hydroxynonenal (4-HNE) modified proteins. Does not detect free 4-Hydroxynonenal. Does not cross-react with 4-Hydroxy-2-hexenal, Acrolein, Crotonaldehyde, Hexanoyl Lysine, Malondialdehyde, or Methylglyoxal modified proteins.
Store at 4C in the dark.