missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenylacetone, 98+%, Thermo Scientific™

Product Code. 11483178
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483178 25 g 25g
11473178 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483178 Supplier Thermo Scientific Alfa Aesar Supplier No. A18936.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 770-39-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00210439
InChI Key VWMVAQHMFFZQGD-UHFFFAOYSA-N
Synonym 4-hydroxyphenylacetone, 1-4-hydroxyphenyl propan-2-one, 1-4-hydroxyphenyl acetone, p-hydroxyphenylacetone, 1-4-hydroxyphenyl-2-propanone, 2-propanone, 1-4-hydroxyphenyl, 4-hydroxyphenyl acetone, 4-hydroxyphenylpropanone, acmc-209p7o, 4-hydroxyphenylpropan-2-one
PubChem CID 7019274
IUPAC Name 1-(4-hydroxyphenyl)propan-2-one
SMILES CC(=O)CC1=CC=C(O)C=C1

Specifications

Quantity 25 g
Sensitivity Air Sensitive
Formula Weight 150.18
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxyphenylacetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.