Learn More
4-Hydroxythiobenzamide, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxythiobenzamide |
| CAS | 25984-63-8 |
| Molecular Formula | C7H7NOS |
| Quantity | 100 g |
| Synonym | p-Hydroxythio-benzamide, 4-Hydroxybenzenecarbothioamide, 4-Hydroxybenzenethiocarboxamide, 4-Hydroxybenzothioamide, 4-Hydroxythiobenzamide, p-Hydroxythiobenzamide |
| InChI Formula | InChI=1S/C7H7NOS/c8-7(10)5-1-3-6(9)4-2-5/h1-4,9H,(H2,8,10) |
| SMILES | NC(=S)c1ccc(O)cc1 |
| IUPAC Name | 4-hydroxybenzenecarbothioamide |
| Molecular Weight (g/mol) | 153.2 |
| Formula Weight | 153.0248 |