Learn More
4-Iodo-N,N-dimethylaniline, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Iodo-N,N-dimethylaniline |
| CAS | 698-70-4 |
| Molecular Formula | C8 H10 I N |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C8H10IN/c1-10(2)8-5-3-7(9)4-6-8/h3-6H,1-2H3 |
| SMILES | CN(C)c1ccc(I)cc1 |
| IUPAC Name | 4-iodo-N,N-dimethyl-aniline |
| Molecular Weight (g/mol) | 247.076 |
| Formula Weight | 246.9858 |