Learn More
4-Iodoaniline, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 4-Iodoaniline |
| CAS | 540-37-4 |
| Molecular Formula | C6H6IN |
| Quantity | 25 g |
| Synonym | 1-Iodo-4-aminobenzene, 4-Iodobenzenamine, 4-Iodobenzeneamine, 4-Iodophenylamine, NSC 9246, p-Aminoiodobenzene, p-Aminophenyl Iodide, p-Iodoaniline |
| InChI Formula | InChI=1S/C6H6IN/c7-5-1-3-6(8)4-2-5/h1-4H,8H2 |
| SMILES | Nc1ccc(I)cc1 |
| IUPAC Name | 4-iodoaniline |
| Molecular Weight (g/mol) | 219.02 |
| Formula Weight | 218.9545 |