Learn More
4-Iodophenylhydrazine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Iodophenylhydrazine Hydrochloride |
| CAS | 62830-55-1 |
| Molecular Formula | C6H7IN2 . (HCl) |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C6H7IN2.ClH/c7-5-1-3-6(9-8)4-2-5;/h1-4,9H,8H2;1H |
| SMILES | NNC1=CC=C(I)C=C1.Cl |
| IUPAC Name | (4-iodophenyl)hydrazine hydrochloride |
| Molecular Weight (g/mol) | 270.5 |
| Formula Weight | 269.95 |