missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Isobutylbenzoic Acid, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
4-Isobutylbenzoic Acid
Specifications
Specifications
| Chemical Name or Material | 4-Isobutylbenzoic Acid |
| CAS | 38861-88-0 |
| Analyte or Component Names | 4-Isobutylbenzoic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H14 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4-Isobutylbenzoic Acid |
| InChI Formula | InChI=1S/C11H14O2/c1-8(2)7-9-3-5-10(6-4-9)11(12)13/h3-6,8H,7H2,1-2H3,(H,12,13) |
| SMILES | CC(C)Cc1ccc(cc1)C(=O)O |
| IUPAC Name | 4-(2-methylpropyl)benzoic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction