missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy 17Beta-Estradiol, TRC

Product Code. 17939444
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17939444 1 mg 1mg
17938114 2.5 mg 2.5mg
17918684 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17939444 Supplier Toronto Research Chemicals Supplier No. TRCM2626301MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26788-23-8
Molecular Formula C19 H26 O3
Molecular Weight (g/mol) 302.41
Synonym Estra-1,3,5(10)-triene-3,17-diol, 4-methoxy-, (17beta)-, Estra-1,3,5(10)-triene-3,17beta-diol, 4-methoxy- (8CI), (17beta)-4-Methoxyestra-1,3,5(10)-triene-3,17-diol, 4-Methoxy-17beta-estradiol, 4-Methoxyestradiol
IUPAC Name (8R,9S,13S,14S,17S)-4-methoxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
SMILES COc1c(O)ccc2[C@H]3CC[C@]4(C)[C@@H](O)CC[C@H]4[C@@H]3CCc12

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C19H26O3/c1-19-10-9-12-11-5-7-16(20)18(22-2)14(11)4-3-13(12)15(19)6-8-17(19)21/h5,7,12-13,15,17,20-21H,3-4,6,8-10H2,1-2H3/t12-,13-,15+,17+,19+/m1/s1
Formula Weight 302.1882
Percent Purity >95%
Chemical Name or Material 4-Methoxy-17 beta-estradiol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.