missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-piperazineacetonitrile, 97%, Thermo Scientific™

Catalog Number p-7055385
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
874-77-1
C7H13N3
139.202
XCUQQNRWWCWVOP-UHFFFAOYSA-N
2-4-methylpiperazin-1-yl acetonitrile, 4-methylpiperazin-1-yl acetonitrile, 4-methyl-1-piperazineacetonitrile, 1-4-methylpiperazine acetonitrile, 1-cyanomethyl-4-methylpiperazine, 1-piperazineacetonitrile, 4-methyl, 4-methyl-piperazin-yl-acetonitrile, 4-methyl-piperazin-1-yl-acetonitrile
10582860
2-(4-methylpiperazin-1-yl)acetonitrile
CN1CCN(CC1)CC#N
This item is not returnable. View return policy
874-77-1
C7H13N3
139.202
XCUQQNRWWCWVOP-UHFFFAOYSA-N
2-4-methylpiperazin-1-yl acetonitrile, 4-methylpiperazin-1-yl acetonitrile, 4-methyl-1-piperazineacetonitrile, 1-4-methylpiperazine acetonitrile, 1-cyanomethyl-4-methylpiperazine, 1-piperazineacetonitrile, 4-methyl, 4-methyl-piperazin-yl-acetonitrile, 4-methyl-piperazin-1-yl-acetonitrile
10582860
2-(4-methylpiperazin-1-yl)acetonitrile
CN1CCN(CC1)CC#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3439
Formula Weight 139.2
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Methyl-1-piperazineacetonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'