missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1H-imidazole-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10801380
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10801380 50 g 50g
10141132 1 g 1g
10203762 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10801380 Supplier Thermo Scientific Maybridge Supplier No. SB01366EE
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 68282-53-1
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00173728
InChI Key KMWCSNCNHSEXIF-UHFFFAOYSA-N
Synonym 4-methyl-1h-imidazole-5-carbaldehyde, 5-methylimidazole-4-carboxaldehyde, 4-methylimidazole-5-carboxaldehyde, 4-methyl-5-imidazolecarboxaldehyde, 5-methyl-3h-imidazole-4-carbaldehyde, 4-methylimidazole-5-carbaldehyde, 1h-imidazole-4-carboxaldehyde, 5-methyl, 1h-imidazole-5-carboxaldehyde, 4-methyl, pubchem23016, acmc-1b6zs
PubChem CID 2795887
IUPAC Name 5-methyl-1H-imidazole-4-carbaldehyde
SMILES CC1=C(C=O)N=CN1

Specifications

Assay Percent Range 97%
Quantity 50 g
Formula Weight 110.12
Percent Purity 97%
Chemical Name or Material 4-Methyl-1H-imidazole-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.