Learn More
4-Methyl-2-oxo-2H-1-benzopyran-7-yl-beta-D-glucopyranosiduronic Acid Methyl Ester 2,3,4-Triacetate, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Methyl-2-oxo-2H-1-benzopyran-7-yl-beta-D-glucopyranosiduronic Acid Methyl Ester 2,3,4-Triacetate |
| CAS | 101014-65-7 |
| Molecular Formula | C23 H24 O12 |
| Quantity | 5 mg |
| Synonym | 4-Methyl-2-oxo-2H-1-benzopyran-7-yl-β-D-glucopyranosiduronic Acid Methyl Ester |
| InChI Formula | InChI=1S/C23H24O12/c1-10-8-17(27)34-16-9-14(6-7-15(10)16)33-23-21(32-13(4)26)19(31-12(3)25)18(30-11(2)24)20(35-23)22(28)29-5/h6-9,18-21,23H,1-5H3/t18-,19-,20-,21+,23+/m0/s1 |
| SMILES | COC(=O)[C@H]1O[C@@H](Oc2ccc3C(=CC(=O)Oc3c2)C)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C |
| IUPAC Name | methyl (2S,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxane-2-carboxylate |
| Molecular Weight (g/mol) | 492.43 |
| Formula Weight | 492.127 |