Learn More
Alfa Aesar™ 4-Methyl-2-phenyl-1,2,3-triazole-5-carboxylic acid, 98%
Product Code. 11377965
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11377965 1 g 1g
11387965 5 g 5g
2 options
Product Code. 11377965
Brand: Alfa Aesar™ L08051.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 22300-56-7
Molecular Formula C10H9N3O2
Molecular Weight (g/mol) 203.201
MDL Number MFCD00051653
InChI Key GQYQHAGPALBYDO-UHFFFAOYSA-N
Synonym 5-methyl-2-phenyl-2h-1,2,3-triazole-4-carboxylic acid, 4-methyl-2-phenyl-1,2,3-triazole-5-carboxylic acid, 5-methyl-2-phenyl-1,2,3-triazole-4-carboxylic acid, 4-methyl-2-phenyl-2h-1,2,3-triazole-5-carboxylic acid, 2h-1,2,3-triazole-4-carboxylic acid, 5-methyl-2-phenyl, acmc-20ame3, maybridge1_002457, 4-carboxy-5-methyl-2-phenyl-1,2,3-triazole, 2-phenyl-5-methyl-4-carboxy-2h-1,2,3-triazole, 2h-1,2,3-triazole-4-carboxylicacid,5-methyl-2-phenyl
PubChem CID 519936
IUPAC Name 5-methyl-2-phenyltriazole-4-carboxylic acid
SMILES CC1=NN(N=C1C(=O)O)C2=CC=CC=C2
Specifications
Melting Point 200°C to 205°C
Quantity 1 g
Beilstein 177649
Formula Weight 203.2
Percent Purity 98%
Chemical Name or Material 4-Methyl-2-phenyl-1,2,3-triazole-5-carboxylic acid