Learn More
4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic Acid |
| CAS | 532391-89-2 |
| Molecular Formula | C10H11NO3 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C10H11NO3/c1-11-4-5-14-9-6-7(10(12)13)2-3-8(9)11/h2-3,6H,4-5H2,1H3,(H,12,13) |
| SMILES | CN1CCOc2cc(ccc12)C(=O)O |
| IUPAC Name | 4-methyl-2,3-dihydro-1,4-benzoxazine-7-carboxylic acid |
| Molecular Weight (g/mol) | 193.2 |
| Formula Weight | 193.0739 |