missing translation for 'onlineSavingsMsg'
Learn More

4-Methylmorpholine N-oxide hydrate, 97%, Thermo Scientific Chemicals

Catalog Number p-165204
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
80913-66-2, 7529-22-8
C5H11NO2
117.15
MFCD00005947
LFTLOKWAGJYHHR-UHFFFAOYSA-N
4-methylmorpholine n-oxide, 4-methylmorpholine 4-oxide, n-methylmorpholine oxide, n-methylmorpholine n-oxide, morpholine, 4-methyl-, 4-oxide, 4-methylmorpholine-4-oxide, methyl morpholine oxide, n-methylmorpholine-n-oxide, nmo, unii-arc64pkj0f
82029
CHEBI:52093
4-methyl-4-oxidomorpholin-4-ium
C[N+]1([O-])CCOCC1
80913-66-2, 7529-22-8
C5H11NO2
117.15
MFCD00005947
LFTLOKWAGJYHHR-UHFFFAOYSA-N
4-methylmorpholine n-oxide, 4-methylmorpholine 4-oxide, n-methylmorpholine oxide, n-methylmorpholine n-oxide, morpholine, 4-methyl-, 4-oxide, 4-methylmorpholine-4-oxide, methyl morpholine oxide, n-methylmorpholine-n-oxide, nmo, unii-arc64pkj0f
82029
CHEBI:52093
4-methyl-4-oxidomorpholin-4-ium
C[N+]1([O-])CCOCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 70°C to 75°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 6171
Formula Weight 117.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Methylmorpholine N-oxide hydrate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methylmorpholine N-oxide hydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'