Learn More
4-Methylpyrimidine, 98%
Product Code. 11414564
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414564 25 g 25g
11404564 5 g 5g
11424564 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11414564 Supplier Thermo Scientific Alfa Aesar Supplier No. A13170.14
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
CAS: 3438-46-8 | C5H6N2 | 94.117 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identificadores de productos químicos
CAS 3438-46-8
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.117
MDL Number MFCD00006115
InChI Key LVILGAOSPDLNRM-UHFFFAOYSA-N
Synonym pyrimidine, 4-methyl, 4-methyl-pyrimidine, 4-methyl pyrimidine, pubchem20803, 6-methylpyrimidine, 4-methylpyrimidine, ksc226a9b, tpc-h013, 4-methyl-1,3-diazine, pyrimidine, 4-methyl-6ci,7ci,8ci,9ci
PubChem CID 18922
IUPAC Name 4-methylpyrimidine
SMILES CC1=NC=NC=C1
Especificaciones
Density 1.03
Boiling Point 141°C to 142°C
Flash Point 40°C (104°F)
Refractive Index 1.496
Quantity 25 g
UN Number UN1993
Beilstein 105781
Formula Weight 94.12
Percent Purity 98%
Chemical Name or Material 4-Methylpyrimidine
RUO – Research Use Only
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.