Learn More
Melford 4-Methylumbelliferyl-α-D-Glucoside
Product Code. 30018516
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
Product Code. 30018516
Brand: Melford M657000.1
Fluorescent substrate for 4-Methylumbelliferyl-α-D-Glucosidase.
Fluorescent substrate for 4-Methylumbelliferyl-α-D-Glucosidase.
4-Methylumbelliferone <50ppm.
Specifications
Quantity 100 mg