missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholinecarbonitrile, TRC

Product Code. 30471779
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471779 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471779 Supplier Toronto Research Chemicals Supplier No. TRCM72391010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Morpholinecarbonitrile
CAS 1530-89-8
Molecular Formula C5H8N2O
Quantity 10 g
Synonym 4-Cyanomorpholine, Morpholine-4-nitrile, Morpholinocarbonitrile, Morpholinocyanamide, N-Cyanomorpholine, N-Morpholinonitrile, NSC 44243
InChI Formula InChI=1S/C5H8N2O/c6-5-7-1-3-8-4-2-7/h1-4H2
SMILES N#CN1CCOCC1
IUPAC Name morpholine-4-carbonitrile
Molecular Weight (g/mol) 112.13
Formula Weight 112.0637
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.