Learn More
4-Nitrophenylacetonitrile, 98%
Product Code. 11422577
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
Product Code. 11422577
Brand: Thermo Scientific Alfa Aesar A14821.18
Request Bulk or Custom Format
CAS: 555-21-5 | C8H6N2O2 | 162.148 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 555-21-5
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00007372
InChI Key PXNJGLAVKOXITN-UHFFFAOYSA-N
Synonym 4-nitrophenylacetonitrile, p-nitrophenylacetonitrile, benzeneacetonitrile, 4-nitro, 4-nitrobenzyl cyanide, 2-4-nitrophenyl acetonitrile, p-nitrobenzyl cyanide, p-nitrobenzylcyanide, 4-nitrobenzeneacetonitrile, acetonitrile, p-nitrophenyl, p-nitrophenyl acetonitrile
PubChem CID 68386
IUPAC Name 2-(4-nitrophenyl)acetonitrile
SMILES C1=CC(=CC=C1CC#N)[N+](=O)[O-]
Specifications
Melting Point 114°C to 117°C
Boiling Point 195°C to 197°C (12 mmHg)
Quantity 50 g
UN Number UN3439
Beilstein 388442
Merck Index 14,6590
Formula Weight 162.15
Percent Purity 98%
Chemical Name or Material 4-Nitrophenylacetonitrile