Learn More
Tocris Bioscience™ 4-P-PDOT
GREENER_CHOICE
Product Code. 18094682
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. 18094682
Brand: Tocris Bioscience™ 1034/50
MT2 antagonist
Melatonin receptor antagonist, >300-fold selective for the MT2 (vs MT1) subtype.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 134865-74-0
Molecular Formula C19H21NO
Specifications
Quantity 50 mg
Formula Weight 279.38
Purity >99%
Molecular Weight (g/mol) 279.383
Chemical Name or Material 4-P-PDOT