missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pentenoic Acid, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 4-Pentenoic Acid |
| CAS | 591-80-0 |
| Molecular Formula | C5H8O2 |
| Quantity | 5 g |
| Synonym | Allylacetic Acid, NSC 20944, NSC 9000, delta4-Pentenoic Acid |
| InChI Formula | InChI=1S/C5H8O2/c1-2-3-4-5(6)7/h2H,1,3-4H2,(H,6,7) |
| SMILES | OC(=O)CCC=C |
| IUPAC Name | pent-4-enoic acid |
| Molecular Weight (g/mol) | 100.12 |
| Formula Weight | 100.0524 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction