Learn More
4-Phenyl-1,2,3,6-tetrahydropyridine hydrochloride, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 43064-12-6 |
| Molecular Formula | C11H14ClN |
| MDL Number | MFCD00012752 |
| Quantity | 1 g |
| SMILES | [H]Cl.C1(C2=CC=CC=C2)=CCNCC1 |
| Molecular Weight (g/mol) | 195.69 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Phenyl-1,2,3,6-tetrahydropyridine hydrochloride |