missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-1-(4-piperidinyl)-4-imidazolin-2-one, 96%, Thermo Scientific™

Catalog Number p-7033677
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
205058-28-2
C14H17N3O
243.31
MFCD08061015
OPCVKWIYGHICKG-UHFFFAOYSA-N
4-phenyl-1-4-piperidinyl-4-imidazolin-2-one, 4-phenyl-1-piperidin-4-yl-1,3-dihydro-2h-imidazol-2-one, 4-phenyl-1-piperidin-4-yl-1h-imidazol-2 3h-one, 4-phenyl-1-piperidin-4-yl-3h-imidazol-2-one, 4-phenyl-1-piperidin-4-yl-2,3-dihydro-1h-imidazol-2-one, 4-phenyl-1-4-piperidyl-4-imidazolin-2-one, acmc-1chii, 4-phenyl-piperidin-4-yl-1h-imidazol-2 3h-one, 1,3-dihydro-1-4-piperidinyl-4-phenyl-2 2h-imidazolone, 2h-imidazol-2-one, 1,3-dihydro-4-phenyl-1-4-piperidinyl
11345546
5-phenyl-3-piperidin-4-yl-1H-imidazol-2-one
C1CNCCC1N2C=C(NC2=O)C3=CC=CC=C3
This item is not returnable. View return policy
205058-28-2
C14H17N3O
243.31
MFCD08061015
OPCVKWIYGHICKG-UHFFFAOYSA-N
4-phenyl-1-4-piperidinyl-4-imidazolin-2-one, 4-phenyl-1-piperidin-4-yl-1,3-dihydro-2h-imidazol-2-one, 4-phenyl-1-piperidin-4-yl-1h-imidazol-2 3h-one, 4-phenyl-1-piperidin-4-yl-3h-imidazol-2-one, 4-phenyl-1-piperidin-4-yl-2,3-dihydro-1h-imidazol-2-one, 4-phenyl-1-4-piperidyl-4-imidazolin-2-one, acmc-1chii, 4-phenyl-piperidin-4-yl-1h-imidazol-2 3h-one, 1,3-dihydro-1-4-piperidinyl-4-phenyl-2 2h-imidazolone, 2h-imidazol-2-one, 1,3-dihydro-4-phenyl-1-4-piperidinyl
11345546
5-phenyl-3-piperidin-4-yl-1H-imidazol-2-one
C1CNCCC1N2C=C(NC2=O)C3=CC=CC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point ∽235°C (decomposition)
Quantity 250 mg
Formula Weight 243.31
Percent Purity 96%
Chemical Name or Material 4-Phenyl-1-(4-piperidinyl)-4-imidazolin-2-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'