missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-sec-Butylbenzeneboronic acid, 98%

Catalog Number 15470098
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
850568-56-8
C10H15BO2
178.038
MFCD02179456
CHSPURIZKQPJNH-UHFFFAOYSA-N
4-sec-butylphenylboronic acid, 4-sec-butylphenyl boronic acid, 4-1-methylpropyl benzeneboronic acid, 4-sec-butyl phenyl boronic acid, 4-1-methylpropyl phenylboronic acid, 4-sec-butyl benzeneboronic acid, 4-sec-butyl phenylboronic acid, 4-butan-2-yl phenyl boronic acid, boronic acid,b-4-1-methylpropyl phenyl, boronic acid, 4-1-methylpropyl phenyl
3690874
(4-butan-2-ylphenyl)boronic acid
B(C1=CC=C(C=C1)C(C)CC)(O)O
This item is not returnable. View return policy
850568-56-8
C10H15BO2
178.038
MFCD02179456
CHSPURIZKQPJNH-UHFFFAOYSA-N
4-sec-butylphenylboronic acid, 4-sec-butylphenyl boronic acid, 4-1-methylpropyl benzeneboronic acid, 4-sec-butyl phenyl boronic acid, 4-1-methylpropyl phenylboronic acid, 4-sec-butyl benzeneboronic acid, 4-sec-butyl phenylboronic acid, 4-butan-2-yl phenyl boronic acid, boronic acid,b-4-1-methylpropyl phenyl, boronic acid, 4-1-methylpropyl phenyl
3690874
(4-butan-2-ylphenyl)boronic acid
B(C1=CC=C(C=C1)C(C)CC)(O)O
Melting Point 96°C to 100°C
Quantity 250 mg
Formula Weight 178.04
Percent Purity 98%
Chemical Name or Material 4-sec-Butylbenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'