missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyldimethylsiloxy)phenol, 97%

Product Code. 15411858
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15411858 5 g 5g
15421858 250 mg 250mg
15401858 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15411858 Supplier Thermo Scientific Alfa Aesar Supplier No. H55556.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 108534-47-0 | C12H20O2Si | 224.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108534-47-0
Molecular Formula C12H20O2Si
Molecular Weight (g/mol) 224.38
MDL Number MFCD06200713
InChI Key KZTVHIZALLBXMO-UHFFFAOYSA-N
Synonym 4-tert-butyldimethylsiloxy phenol, 4-tert-butyldimethylsilyl oxy phenol, 4-t-butyldimethylsiloxy phenol, 4-tert-butyldimethylsilyloxy phenol, 4-tert-butyl dimethyl silyl oxyphenol, phenol, 4-t-butyldimethylsilyloxy, acmc-20amtw, hydroquinone, tbdms derivative, 4-tert-butyldimethylsilyloxyphenol
PubChem CID 597388
IUPAC Name 4-[tert-butyl(dimethyl)silyl]oxyphenol
SMILES CC(C)(C)[Si](C)(C)OC1=CC=C(O)C=C1

Specifications

Melting Point 60°C to 64°C
Color White
Quantity 5 g
Formula Weight 224.38
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-(tert-Butyldimethylsiloxy)phenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.