Learn More
4-(tert-Butyldimethylsilyloxymethyl)bromobenzene, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(tert-Butyldimethylsilyloxymethyl)bromobenzene |
| CAS | 87736-74-1 |
| Molecular Formula | C13H21BrOSi |
| Quantity | 1 g |
| Synonym | 4-Bromobenzyl dimethyl-tert-butylsilyl Ether, (4-Bromobenzyl)oxy](tert-butyl)dimethylsilane, 1-Bromo-4-[[[(1,1-dimethylethyl)dimethylsilyl]oxy]methyl]-benzene |
| InChI Formula | InChI=1S/C13H21BrOSi/c1-13(2,3)16(4,5)15-10-11-6-8-12(14)9-7-11/h6-9H,10H2,1-5H3 |
| SMILES | CC(C)(C)[Si](C)(C)OCc1ccc(Br)cc1 |
| IUPAC Name | (4-bromophenyl)methoxy-tert-butyl-dimethylsilane |
| Molecular Weight (g/mol) | 301.29 |
| Formula Weight | 300.0545 |