Learn More
4-(Trifluoromethyl)pyridin-3-amine, 97%, Thermo Scientific™
Product Code. 10754583
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10754583 1 g 1g
10516144 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10754583 Supplier Thermo Scientific Maybridge Supplier No. RF03857DA
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Chemical Identifiers
CAS 175204-80-5
Molecular Formula C6H5F3N2
Molecular Weight (g/mol) 162.12
MDL Number MFCD00173960
InChI Key DROFUWWORPKKSI-UHFFFAOYSA-N
Synonym 3-amino-4-trifluoromethyl pyridine, 4-trifluoromethyl pyridin-3-amine, 3-amino-4-trifluoromethylpyridine, 4-trifluoromethyl-pyridin-3-ylamine, 3-pyridinamine, 4-trifluoromethyl, 4-trifluoromethyl-3-pyridinamine, 4-trifluoromethyl-3-pyridylamine, pubchem2987, acmc-209e9a, 4-trifluoromethyl-3-pyridinylamine
PubChem CID 2735957
IUPAC Name 4-(trifluoromethyl)pyridin-3-amine
SMILES NC1=C(C=CN=C1)C(F)(F)F
Specifications
CAS Min % 97.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 162.11
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)pyridin-3-amine