Learn More
4-Vinylphenol, 95%, 10% solution in propylene glycol, Thermo Scientific Chemicals
Catalog Number p-4258825
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2628-17-3, 57-55-6
C8H8O
120.16
FUGYGGDSWSUORM-UHFFFAOYSA-N
4-vinylphenol, 4-hydroxystyrene, p-vinylphenol, p-hydroxystyrene, phenol, 4-ethenyl, 4-vinylphenol, 10 wt.% in propylene glycol, phenol, p-vinyl, unii-oa7v1sm8yl, fema no. 3739, poly 4-hydroxystyrene
62453
CHEBI:1883
4-ethenylphenol
C=CC1=CC=C(C=C1)O
2628-17-3, 57-55-6
C8H8O
120.16
FUGYGGDSWSUORM-UHFFFAOYSA-N
4-vinylphenol, 4-hydroxystyrene, p-vinylphenol, p-hydroxystyrene, phenol, 4-ethenyl, 4-vinylphenol, 10 wt.% in propylene glycol, phenol, p-vinyl, unii-oa7v1sm8yl, fema no. 3739, poly 4-hydroxystyrene
62453
CHEBI:1883
4-ethenylphenol
C=CC1=CC=C(C=C1)O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 89.0, 9.0
CAS Max % 91.0, 11.0
Color Colorless to Orange
Density 1.0390g/mL
Boiling Point 189°C
Infrared Spectrum Authentic
Assay Percent Range 9 to 15% (GC)
Packaging Glass bottle
Refractive Index 1.4420 to 1.4520 (20°C, 589nm)
Quantity 25 mL
Specific Gravity 1.039
Formula Weight 120.16
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Vinylphenol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Vinylphenol, 10% solution in propylene glycol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.