Learn More
Schneider Gerd™ 4301 Stainless Steel Micro Spoon Spatula
Use this spatula for a variety of applications. Schneider Gerd™ 4301 Stainless Steel Micro Spoon Spatulas are ideal for use with a wide range of compounds.
Brand: Schneider Gerd™ 253
Additional Details : Weight : 0.10000kg
Description
- Stainless steel micro spatula
- Spoons collect and transport dry substances or liquids
- Ideal for working with powders, stirring and mixing
- Perfect for miniscule amounts of chemicals
Specifications
Micro Spoon Spatula | |
6 mm | |
Stainless Steel (4301) | |
185 mm |