Learn More
Schneider Gerd™ 4301 Stainless Steel Micro Spoon Spatula
Use this spatula for a variety of applications. Schneider Gerd™ 4301 Stainless Steel Micro Spoon Spatulas are ideal for use with a wide range of compounds.
Brand:  Schneider Gerd™ 253
Additional Details : Weight : 0.10000kg
Product Code. 10071654
  • € 7.00 / Each
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
Description
Description
  • Stainless steel micro spatula
  • Spoons collect and transport dry substances or liquids
  • Ideal for working with powders, stirring and mixing
  • Perfect for miniscule amounts of chemicals
Specifications
Specifications
Micro Spoon Spatula
6 mm
Stainless Steel (4301)
185 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers