Learn More
Schneider Gerd™ 4301 Stainless Steel Spoon
€ 7.00 - € 11.00
Specifications
For Use With (Application) | For Chemical |
---|---|
Material | Stainless Steel (4301) |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10006961
|
Schneider Gerd™
243 |
180 mm |
€ 7.70
Each |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
10672835
|
Schneider Gerd™
246 |
300 mm |
€ 11.00
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
10326604
|
Schneider Gerd™
245 |
250 mm |
€ 10.00
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
10011424
|
Schneider Gerd™
244 |
210 mm |
€ 7.00
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
10001424
|
Schneider Gerd™
242 |
150 mm |
€ 7.00
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Specifications
For Chemical | |
Stainless Steel (4301) |