Learn More
Hamilton™ Gastight Microliter™ Syringes: RN Termination, No Needle Included
Click to view available options
Volume (Metric):
5 μL
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
1 mL
2.5 mL
5 mL
10 mL
Model:
1001
1002
1005
1010
1725
1750
1801 RN
1802
1805
1810 RN
1825
701
702
705
710
725
750
75RN
missing translation for 'unitSize'
Each
Description
- ideal for dispensing volumes from 0.05 µL up to 5 µL
- The 7000 series is the only zero dead volume syringe
- The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle
Specifications
Specifications
| Description | 710RN |
| Needle Gauge | 22 G |
| Model | 710 |
| Volume (Metric) | 100 μL |