missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4EBP1 Antibody (0014) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 160.00 - € 428.00
Specifications
| Antigen | 4EBP1 |
|---|---|
| Clone | 0014 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18646654
|
Novus Biologicals
NBP3-23518-20ul |
20 μL |
€ 160.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18680724
|
Novus Biologicals
NBP3-23518-100ul |
100 μL |
€ 428.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4EBP1 Monoclonal antibody specifically detects 4EBP1 in Human samples. It is validated for Western BlotSpecifications
| 4EBP1 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 4E-BP1, eIF4E-binding protein 1, eukaryotic translation initiation factor 4E binding protein 1,4EBP1,4E-BP1BP-1, eukaryotic translation initiation factor 4E-binding protein 1, PHAS-IMGC4316, Phosphorylated heat- and acid-stable protein regulated by insulin 1 | |
| This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to residues around (Thr37, 46) of Human phospho-4EBP1 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0014 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Lipid and Metabolism, mTOR Pathway, Phospho Specific, Stem Cell Markers | |
| 0.2 um filtered solution in PBS | |
| 1978 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title