Learn More
(4S)-2-[[[(2R)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]methyl]-5,5-dimethyl-4-thiazolidinecarboxylic Acid (Mixture of Diastereomers), TRC
Specifications
Specifications
| Chemical Name or Material | (4S)-2-[([(2R)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino)methyl]-5,5-dimethyl-4-thiazolidinecarboxylic Acid (Mixture of Diastereomers) |
| CAS | 1356020-01-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C15 H21 N3 O4 S |
| Quantity | 50 mg |
| Synonym | Amoxicillin EP Impurity E, Amoxicillin USP Related Compound E |
| InChI Formula | InChI=1S/C15H21N3O4S/c1-15(2)12(14(21)22)18-10(23-15)7-17-13(20)11(16)8-3-5-9(19)6-4-8/h3-6,10-12,18-19H,7,16H2,1-2H3,(H,17,20)(H,21,22)/t10?,11-,12+/m1/s1 |
| SMILES | CC1([C@@H](NC(S1)CNC(=O)[C@@H](C2=CC=C(C=C2)O)N)C(=O)O)C |
| IUPAC Name | (4S)-2-(((R)-2-Amino-2-(4-hydroxyphenyl)acetamido)methyl)-5,5-dimethylthiazolidine-4-carboxylic Acid |
| Molecular Weight (g/mol) | 339.41 |
| Show More |