Learn More
5-(1-Pyrrolidinyl)-3(2H)-pyridazinone, TRC
Specifications
Specifications
| Chemical Name or Material | 5-(1-Pyrrolidinyl)-3(2H)-pyridazinone |
| CAS | 1015846-81-7 |
| Molecular Formula | C8H11N3O |
| Quantity | 100 mg |
| Synonym | 5-(1-Pyrrolidinyl)-3-pyridazinol |
| InChI Formula | InChI=1S/C8H11N3O/c12-8-5-7(6-9-10-8)11-3-1-2-4-11/h5-6H,1-4H2,(H,10,12) |
| SMILES | O=C1NN=CC(=C1)N2CCCC2 |
| IUPAC Name | 4-pyrrolidin-1-yl-1H-pyridazin-6-one |
| Molecular Weight (g/mol) | 165.19 |
| Formula Weight | 165.0902 |