missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(2-Chlorobenzoyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
5-(2-Chlorobenzoyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine
Specifications
Specifications
| Chemical Name or Material | 5-(2-Chlorobenzoyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine |
| CAS | 68559-48-8 |
| Analyte or Component Names | 5-(2-Chlorobenzoyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H12 Cl N O S |
| Certifications/Compliance | ISO 9001 |
| Synonym | 5-(2-Chlorobenzoyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine |
| InChI Formula | InChI=1S/C14H12ClNOS/c15-12-4-2-1-3-11(12)14(17)16-7-5-13-10(9-16)6-8-18-13/h1-4,6,8H,5,7,9H2 |
| SMILES | Clc1ccccc1C(=O)N2CCc3sccc3C2 |
| IUPAC Name | (2-chlorophenyl)-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)methanone |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction